3-Bromo-5-(dimethoxymethyl)pyridine
Empirical Formula (Hill Notation):
C8H10BrNO2
CAS Number:
Molecular Weight:
232.07
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing