![3-Bromo-4-chloro-thieno[3,2-c]pyridine 95%](/deepweb/assets/sigmaaldrich/product/structures/162/901/f3398048-dc5e-42a8-a005-07307b6e5c8d/640/f3398048-dc5e-42a8-a005-07307b6e5c8d.png)
3-Bromo-4-chloro-thieno[3,2-c]pyridine
![3-Bromo-4-chloro-thieno[3,2-c]pyridine 95%](/deepweb/assets/sigmaaldrich/product/structures/162/901/f3398048-dc5e-42a8-a005-07307b6e5c8d/640/f3398048-dc5e-42a8-a005-07307b6e5c8d.png)
Empirical Formula (Hill Notation):
C7H3BrClNS
CAS Number:
Molecular Weight:
248.53
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing