Skip to Content
Merck
3-Amino-2-bromo-5-fluoropyridine 97%

3-Amino-2-bromo-5-fluoropyridine

3-Amino-2-bromo-5-fluoropyridine 97%
Synonym(s):
2-Bromo-5-fluoro-pyridin-3-amine
Empirical Formula (Hill Notation):
C5H4BrFN2
Molecular Weight:
191.00

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing