Skip to Content
Merck
3,4-O-Isopropylidene-D-mannitol 97%

3,4-O-Isopropylidene-D-mannitol

3,4-O-Isopropylidene-D-mannitol 97%
Empirical Formula (Hill Notation):
C9H18O6
CAS Number:
Molecular Weight:
222.24

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing