Skip to Content
Merck
3-(4-methyl-piperazin-1-ylmethyl)-benzoic acid 95%

3-(4-methyl-piperazin-1-ylmethyl)-benzoic acid

3-(4-methyl-piperazin-1-ylmethyl)-benzoic acid 95%
Empirical Formula (Hill Notation):
C13H18N2O2
CAS Number:
Molecular Weight:
234.29

Products

Brand
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing