Skip to Content
Merck
3,4-Dimethylphenylhydrazine hydrochloride 97%

3,4-Dimethylphenylhydrazine hydrochloride

3,4-Dimethylphenylhydrazine hydrochloride 97%
Linear Formula:
(CH3)2C6H3NHNH2·HCl
CAS Number:
Molecular Weight:
172.66

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing