Skip to Content
Merck
3-(3,4-Dimethoxyphenyl)propionic acid 99%

3-(3,4-Dimethoxyphenyl)propionic acid

3-(3,4-Dimethoxyphenyl)propionic acid 99%
Synonym(s):
3,4-Dimethoxyhydrocinnamic acid
Linear Formula:
(CH3O)2C6H3CH2CH2CO2H
CAS Number:
Molecular Weight:
210.23
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing