![2-(Trifluoromethyl)pyridine 97%](/deepweb/assets/sigmaaldrich/product/structures/289/508/4826157b-4d42-4292-967d-fdad2ae6b2f7/640/4826157b-4d42-4292-967d-fdad2ae6b2f7.png)
2-(Trifluoromethyl)pyridine
![2-(Trifluoromethyl)pyridine 97%](/deepweb/assets/sigmaaldrich/product/structures/289/508/4826157b-4d42-4292-967d-fdad2ae6b2f7/640/4826157b-4d42-4292-967d-fdad2ae6b2f7.png)
Synonym(s):
α,α,α-Trifluoro-2-picoline
Empirical Formula (Hill Notation):
C6H4F3N
CAS Number:
Molecular Weight:
147.10
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing