Skip to Content
Merck
2-(Methylthio)pyrazine FG

2-(Methylthio)pyrazine

2-(Methylthio)pyrazine FG
Synonym(s):
Methyl pyrazinyl sulfide
Empirical Formula (Hill Notation):
C5H6N2S
CAS Number:
Molecular Weight:
126.18
EC Number:

Products

Product Category
Brand
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Agency
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing