Skip to Content
Merck
2-Methylpyridine-3-carboxaldehyde 97%

2-Methylpyridine-3-carboxaldehyde

2-Methylpyridine-3-carboxaldehyde 97%
Empirical Formula (Hill Notation):
C7H7NO
CAS Number:
Molecular Weight:
121.14

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing