Skip to Content
Merck
2-Methyl-6-nitrobenzoic acid 97%

2-Methyl-6-nitrobenzoic acid

2-Methyl-6-nitrobenzoic acid 97%
Synonym(s):
6-Nitro-o-toluic acid
Linear Formula:
CH3C6H3(NO2)CO2H
CAS Number:
Molecular Weight:
181.15
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing