2-Methyl-2-propene-1-sulfonic acid sodium salt
Linear Formula:
H2C=C(CH3)CH2SO3Na
CAS Number:
Molecular Weight:
158.15
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing