Skip to Content
Merck
2-Methyl-1,5-hexadiene 97%

2-Methyl-1,5-hexadiene

2-Methyl-1,5-hexadiene 97%
Synonym(s):
2-Methyldiallyl
Linear Formula:
CH2=CHCH2CH2C(CH3)=CH2
CAS Number:
Molecular Weight:
96.17

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing