Skip to Content
Merck
2-Methoxy-5-nitrophenol 98%

2-Methoxy-5-nitrophenol

2-Methoxy-5-nitrophenol 98%
Synonym(s):
5-Nitroguaiacol
Linear Formula:
CH3OC6H3(NO2)OH
CAS Number:
Molecular Weight:
169.13
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing