Skip to Content
Merck
2-Methoxy-4-methylbenzoic acid 97%

2-Methoxy-4-methylbenzoic acid

2-Methoxy-4-methylbenzoic acid 97%
Empirical Formula (Hill Notation):
C9H10O3
CAS Number:
Molecular Weight:
166.17

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing