Skip to Content
Merck
2-Fluoro-4-methylaniline 99%

2-Fluoro-4-methylaniline

2-Fluoro-4-methylaniline 99%
Synonym(s):
2-Fluoro-p-toluidine
Linear Formula:
FC6H3(CH3)NH2
CAS Number:
Molecular Weight:
125.14

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing