
2-Ethylbutyl acetate

Linear Formula:
CH3CO2CH2CH(C2H5)2
Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Application
Markush Class
Markush Group
Organoleptic Class
Showing 1-1 of 1
Compare
Product Number
Description
Pricing