Skip to Content
Merck
2-Chloro-6-fluorophenylacetonitrile 98%

2-Chloro-6-fluorophenylacetonitrile

2-Chloro-6-fluorophenylacetonitrile 98%
Synonym(s):
2-Chloro-6-fluorobenzyl cyanide
Linear Formula:
ClC6H3(F)CH2CN
CAS Number:
Molecular Weight:
169.58
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing