Skip to Content
Merck
2-Chloro-6-fluorobenzyl chloride 98%

2-Chloro-6-fluorobenzyl chloride

2-Chloro-6-fluorobenzyl chloride 98%
Synonym(s):
2,α-Dichloro-6-fluorotoluene
Linear Formula:
ClC6H3(F)CH2Cl
CAS Number:
Molecular Weight:
179.02
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing