Skip to Content
Merck
2-Chloro-5-fluoropyrimidine 97%

2-Chloro-5-fluoropyrimidine

2-Chloro-5-fluoropyrimidine 97%
Empirical Formula (Hill Notation):
C4H2ClFN2
CAS Number:
Molecular Weight:
132.52

Products

Product Category
Brand
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing