Skip to Content
Merck
2-Chloro-5-fluorobenzylamine 95%

2-Chloro-5-fluorobenzylamine

2-Chloro-5-fluorobenzylamine 95%
Synonym(s):
(2-Chloro-5-fluorophenyl)methanamine, 2-Chloro-5-fluorobenzenemethanamine
Empirical Formula (Hill Notation):
C7H7ClFN
CAS Number:
Molecular Weight:
159.59

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing