Skip to Content
Merck
2-Chloro-4-hydroxybenzaldehyde 97%

2-Chloro-4-hydroxybenzaldehyde

2-Chloro-4-hydroxybenzaldehyde 97%
Linear Formula:
ClC6H3(OH)CHO
CAS Number:
Molecular Weight:
156.57

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing