
2-Chloro-4-benzamido-5-methylbenzenediazonium chloride hemi(zinc chloride) salt

Linear Formula:
C14H11Cl2N3O · 1/2ZnCl2
CAS Number:
Molecular Weight:
376.31
Products
Product Category
Brand
Formula Weight
Physical Form
Functional Group
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing