Skip to Content
Merck
2-Chloro-4,5-difluorobenzoic acid 99%

2-Chloro-4,5-difluorobenzoic acid

2-Chloro-4,5-difluorobenzoic acid 99%
Linear Formula:
ClC6H2(F)2CO2H
CAS Number:
Molecular Weight:
192.55

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing