Skip to Content
Merck
2-Chloro-1,3-dimethylimidazolidinium tetrafluoroborate ≥95.0% (AT)

2-Chloro-1,3-dimethylimidazolidinium tetrafluoroborate

2-Chloro-1,3-dimethylimidazolidinium tetrafluoroborate ≥95.0% (AT)
Synonym(s):
CIB
Empirical Formula (Hill Notation):
C5H10BClF4N2
CAS Number:
Molecular Weight:
220.40

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing