2-Bromo-5-methoxybenzyl bromide
Linear Formula:
BrC6H3(CH2Br)OCH3
CAS Number:
Molecular Weight:
279.96
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing