Skip to Content
Merck
2-Bromo-4-fluorobenzoic acid 97%

2-Bromo-4-fluorobenzoic acid

2-Bromo-4-fluorobenzoic acid 97%
Linear Formula:
BrC6H3(F)CO2H
CAS Number:
Molecular Weight:
219.01

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing