
2-Arachidonoyl-1-palmitoyl-sn-glycero-3-phosphocholine

Synonym(s):
1-Hexadecanoyl-2-([cis,cis,cis,cis]-5,8,11,14-eicosatetraenoyl)-sn-glycero-3-phosphocholine, L-α-Phosphatidylcholine, β-arachidonoyl-γ-palmitoyl
Empirical Formula (Hill Notation):
C44H80NO8P
CAS Number:
Molecular Weight:
782.08
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Functional Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing