Skip to Content
Merck
2-Amino-6-fluorobenzonitrile 99%

2-Amino-6-fluorobenzonitrile

2-Amino-6-fluorobenzonitrile 99%
Synonym(s):
6-Fluoroanthranilonitrile
Linear Formula:
H2NC6H3(F)CN
CAS Number:
Molecular Weight:
136.13

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing