
2-Amino-5-bromo-3-methoxypyrazine

Synonym(s):
3-Bromo-5-methoxy-2-pyrazinamine
Empirical Formula (Hill Notation):
C5H6BrN3O
CAS Number:
Molecular Weight:
204.02
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing