Skip to Content
Merck
2-Amino-4-methoxy-6-methylpyrimidine 98%

2-Amino-4-methoxy-6-methylpyrimidine

2-Amino-4-methoxy-6-methylpyrimidine 98%
Empirical Formula (Hill Notation):
C6H9N3O
CAS Number:
Molecular Weight:
139.16

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing