Skip to Content
Merck
2-Acetylbenzothiophene 97%

2-Acetylbenzothiophene

2-Acetylbenzothiophene 97%
Synonym(s):
1-Benzo[b]thiophen-2-yl-ethanone
Empirical Formula (Hill Notation):
C10H8OS
CAS Number:
Molecular Weight:
176.23
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing