Skip to Content
Merck
2-Acetylamino-5-bromopyridine 98%

2-Acetylamino-5-bromopyridine

2-Acetylamino-5-bromopyridine 98%
Empirical Formula (Hill Notation):
C7H7BrN2O
CAS Number:
Molecular Weight:
215.05

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing