Skip to Content
Merck
2-Acetyl-4-methylthiophene 97%

2-Acetyl-4-methylthiophene

2-Acetyl-4-methylthiophene 97%
Synonym(s):
1-(4-Methyl-2-thienyl)ethanone
Empirical Formula (Hill Notation):
C7H8OS
CAS Number:
Molecular Weight:
140.20
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing