Skip to Content
Merck
2-Acetyl-3,5(6)-dimethylpyrazine, mixture of isomers ≥98%, FG

2-Acetyl-3,5(6)-dimethylpyrazine, mixture of isomers

2-Acetyl-3,5(6)-dimethylpyrazine, mixture of isomers ≥98%, FG
Empirical Formula (Hill Notation):
C8H10N2O
CAS Number:
Molecular Weight:
150.18
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing