
2,6-Diisopropyl-N,N-dimethylaniline

Linear Formula:
[(CH3)2CH]2C6H3N(CH3)2
CAS Number:
Molecular Weight:
205.34
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing