Skip to Content
Merck
2,6-Dichloro-3-nitropyridine technical grade, 92%

2,6-Dichloro-3-nitropyridine

2,6-Dichloro-3-nitropyridine technical grade, 92%
Empirical Formula (Hill Notation):
C5H2Cl2N2O2
CAS Number:
Molecular Weight:
192.99
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing