
2,5-Dichloro-p-phenylenediamine

Synonym(s):
1,4-Diamino-2,5-dichlorobenzene
Linear Formula:
Cl2C6H2(NH2)2
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing