Skip to Content
Merck
2-(4-Formylphenyl)-6-(hydroxymethyl)pyridine 95%

2-(4-Formylphenyl)-6-(hydroxymethyl)pyridine

2-(4-Formylphenyl)-6-(hydroxymethyl)pyridine 95%
Empirical Formula (Hill Notation):
C13H11NO2
CAS Number:
Molecular Weight:
213.23

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing