Skip to Content
Merck
2,4-Dinitrobenzenesulfonyl chloride 97%

2,4-Dinitrobenzenesulfonyl chloride

2,4-Dinitrobenzenesulfonyl chloride 97%
Linear Formula:
(O2N)2C6H3SO2Cl
CAS Number:
Molecular Weight:
266.62

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing