Skip to Content
Merck
2,4,5-Trichlorobenzenesulfonyl chloride 95%

2,4,5-Trichlorobenzenesulfonyl chloride

2,4,5-Trichlorobenzenesulfonyl chloride 95%
Empirical Formula (Hill Notation):
C6H2Cl4O2S
CAS Number:
Molecular Weight:
279.96
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing