2,3-Naphthalenedicarbonitrile
Linear Formula:
C10H6(CN)2
CAS Number:
Molecular Weight:
178.19
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing