Skip to Content
Merck
2-(3-Methoxyphenyl)cyclohexanone technical grade, 90%

2-(3-Methoxyphenyl)cyclohexanone

2-(3-Methoxyphenyl)cyclohexanone technical grade, 90%
Linear Formula:
CH3OC6H4C6H9(=O)
CAS Number:
Molecular Weight:
204.26
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing