Skip to Content
Merck
2′,3′-Dideoxyuridine 98%

2′,3′-Dideoxyuridine

2′,3′-Dideoxyuridine 98%
Synonym(s):
ddU
Empirical Formula (Hill Notation):
C9H12N2O4
CAS Number:
Molecular Weight:
212.20

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing