2,3-Dichlorophenoxyacetic acid
Synonym(s):
2,3-D
Empirical Formula (Hill Notation):
C8H6Cl2O3
CAS Number:
Molecular Weight:
221.04
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Product Line
Purity
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing