Skip to Content
Merck
2,3,5,6-Tetrafluoro-4-(trifluoromethyl)benzyl bromide 98%

2,3,5,6-Tetrafluoro-4-(trifluoromethyl)benzyl bromide

2,3,5,6-Tetrafluoro-4-(trifluoromethyl)benzyl bromide 98%
Synonym(s):
1-(Bromomethyl)-2,3,5,6-tetrafluoro-4-(trifluoromethyl)benzene, 4-(Trifluoromethyl)-2,3,5,6-tetrafluorobenzyl bromide
Linear Formula:
CF3C6F4CH2Br
CAS Number:
Molecular Weight:
310.99

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing