Skip to Content
Merck
2,3,5,6-Tetrabromo-p-xylene analytical standard

2,3,5,6-Tetrabromo-p-xylene

2,3,5,6-Tetrabromo-p-xylene analytical standard
Synonym(s):
1,2,4,5-Tetrabromo-3,6-dimethylbenzene
Linear Formula:
C6Br4(CH3)2
CAS Number:
Molecular Weight:
421.75
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing