Skip to Content
Merck
2,3,4-Trichloroanisole PESTANAL®, analytical standard

2,3,4-Trichloroanisole

2,3,4-Trichloroanisole PESTANAL®, analytical standard
Empirical Formula (Hill Notation):
C7H5Cl3O
CAS Number:
Molecular Weight:
211.47
EC Number:

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing