Skip to Content
Merck
2,2-Dimethyl-3,6,9,12-tetraoxa-2-silatridecane 97%, anhydrous

2,2-Dimethyl-3,6,9,12-tetraoxa-2-silatridecane

2,2-Dimethyl-3,6,9,12-tetraoxa-2-silatridecane 97%, anhydrous
Empirical Formula (Hill Notation):
C10H24O4Si
CAS Number:
Molecular Weight:
236.38

Products

Brand
Formula Weight
Physical Form
Purity
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing