![2-(2-Chlorophenyl)benzimidazole 97%](/deepweb/assets/sigmaaldrich/product/structures/389/464/f49ca667-8441-4fc0-853f-6f67f2d6d93e/640/f49ca667-8441-4fc0-853f-6f67f2d6d93e.png)
2-(2-Chlorophenyl)benzimidazole
![2-(2-Chlorophenyl)benzimidazole 97%](/deepweb/assets/sigmaaldrich/product/structures/389/464/f49ca667-8441-4fc0-853f-6f67f2d6d93e/640/f49ca667-8441-4fc0-853f-6f67f2d6d93e.png)
Empirical Formula (Hill Notation):
C13H9ClN2
CAS Number:
Molecular Weight:
228.68
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing